Ethyl 4-bromo-2-fluorobenzoate
Lieferant: Apollo Scientific
|
Achtung
|
PC53347-1G
PC53347-25G
PC53347-5G
APOSPC53347-1GEA
21.9
EUR
APOSPC53347-1G
APOSPC53347-25G
APOSPC53347-5G
Ethyl 4-bromo-2-fluorobenzoate
Ethyl 4-bromo-2-fluorobenzoate
Formel:
C₉H₈BrFO₂ Molecular Weight: 247,061 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06204444 CAS-Nummer: 474709-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...