1-Bromo-3-tert-butyl-5-fluorobenzene 95+%

Lieferant: Apollo Scientific
Achtung

PC53360-100MG PC53360-1G PC53360-250MG
APOSPC53360-100MGEA 132 EUR
APOSPC53360-100MG APOSPC53360-1G APOSPC53360-250MG
1-Bromo-3-tert-butyl-5-fluorobenzene 95+%
1-Bromo-3-tert-butyl-5-fluorobenzene
Formel: C₁₀H₁₂BrF
Molecular Weight: 231,105 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13193246
CAS-Nummer: 1123172-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR