2-Fluoro-1-iodo-4-(trifluoromethoxy)benzene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Fluoro-4-iodophenyl trifluoromethyl ether
PC53370-250MG
PC53370-500MG
PC53370-5G
APOSPC53370-250MGEA
53.8
EUR
APOSPC53370-250MG
APOSPC53370-500MG
APOSPC53370-5G
2-Fluoro-1-iodo-4-(trifluoromethoxy)benzene
2-Fluoro-1-iodo-4-(trifluoromethoxy)benzene
Formel:
C₇H₃F₄IO Molecular Weight: 305,996 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20526760 CAS-Nummer: 1321963-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...