2-[6-(Trifluoromethyl)pyridin-3-yl]acetic acid 95+%
Lieferant: Apollo Scientific
Synonyme:
2-(6-(Trifluoromethyl)pyridin-3-yl)acetic acid
PC53398-100MG
PC53398-1G
PC53398-250MG
APOSPC53398-100MGEA
154
EUR
APOSPC53398-100MG
APOSPC53398-1G
APOSPC53398-250MG
2-[6-(Trifluoromethyl)pyridin-3-yl]acetic acid 95+%
2-[6-(Trifluoromethyl)pyridin-3-yl]acetic acid
Formel:
C₈H₆F₃NO₂ Molecular Weight: 205.134 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09924985 CAS-Nummer: 913839-73-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...