Methyl 2,4-difluoro-5-iodobenzoate

Lieferant: Apollo Scientific
Achtung

PC53400-1G PC53400-250MG PC53400-5G
APOSPC53400-1GEA 406 EUR
APOSPC53400-1G APOSPC53400-250MG APOSPC53400-5G
Methyl 2,4-difluoro-5-iodobenzoate
Methyl 2,4-difluoro-5-iodobenzoate
Formel: C₈H₅F₂IO₂
Molecular Weight: 298,025 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24107099
CAS-Nummer: 1113050-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR