4-Fluoro-N-piperidin-4-ylbenzamide

Lieferant: Apollo Scientific
Achtung

PC53414-1G PC53414-250MG
APOSPC53414-1GEA 611 EUR
APOSPC53414-1G APOSPC53414-250MG
4-Fluoro-N-piperidin-4-ylbenzamide
4-Fluoro-N-piperidin-4-ylbenzamide
Formel: C₁₂H₁₅FN₂O
Molecular Weight: 222,259 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06740661
CAS-Nummer: 75484-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR