4-Amino-3-bromo-5-nitrophenylsulphur pentafluoride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Amino-3-bromo-5-(pentafluorosulphanyl)nitrobenzene, 2-Bromo-6-nitro-4-(pentafluorothio)aniline
PC5343-1G
PC5343-250MG
APOSPC5343-1GEA
973
EUR
APOSPC5343-1G
APOSPC5343-250MG
4-Amino-3-bromo-5-nitrophenylsulphur pentafluoride
4-Amino-3-bromo-5-nitrophenylsulphur pentafluoride
Formel:
C₆H₄BrF₅N₂O₂S Molecular Weight: 343,069 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11501049 CAS-Nummer: 1159512-33-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...