5-Bromo-2-(trifluoromethyl)benzyl acetate

Lieferant: Apollo Scientific
Achtung

PC53435-1G PC53435-250MG
APOSPC53435-1GEA 201 EUR
APOSPC53435-1G APOSPC53435-250MG
5-Bromo-2-(trifluoromethyl)benzyl acetate
5-Bromo-2-(trifluoromethyl)benzyl acetate
Formel: C₁₀H₈BrF₃O₂
Molecular Weight: 297,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30481008
CAS-Nummer: 2149591-27-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR