6-Bromo-8-(trifluoromethoxy)-1,2,3,4-tetrahydroquinolin-2-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-8-(trifluoromethoxy)-3,4-dihydroquinolin-2(1H)-one
PC53440-1G
PC53440-250MG
APOSPC53440-1GEA
140
EUR
APOSPC53440-1G
APOSPC53440-250MG
6-Bromo-8-(trifluoromethoxy)-1,2,3,4-tetrahydroquinolin-2-one
6-Bromo-8-(trifluoromethoxy)-1,2,3,4-tetrahydroquinolin-2-one
Formel:
C₁₀H₇BrF₃NO₂ Molecular Weight: 310,067 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30492324 CAS-Nummer: 2149591-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...