2-Fluoro-3-iodonitrobenzene

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-1-iodo-3-nitrobenzene

PC53450-1G PC53450-250MG PC53450-5G
APOSPC53450-1GEA 493 EUR
APOSPC53450-1G APOSPC53450-250MG APOSPC53450-5G
2-Fluoro-3-iodonitrobenzene
2-Fluoro-3-iodonitrobenzene
Formel: C₆H₃FINO₂
Molecular Weight: 266,996 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18396988
CAS-Nummer: 1261782-23-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR