1-Bromo-3-[(trifluoromethyl)sulphonyl]benzene

Lieferant: Apollo Scientific
Achtung

PC53462-1G PC53462-250MG
APOSPC53462-1GEA 400 EUR
APOSPC53462-1G APOSPC53462-250MG
1-Bromo-3-[(trifluoromethyl)sulphonyl]benzene
1-Bromo-3-[(trifluoromethyl)sulphonyl]benzene
Formel: C₇H₄BrF₃O₂S
Molecular Weight: 289.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28134183
CAS-Nummer: 2728-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR