2,3-Difluoro-4-formylbenzonitrile 96%

Lieferant: Apollo Scientific
Achtung

PC53463-1G PC53463-250MG
APOSPC53463-1GEA 906 EUR
APOSPC53463-1G APOSPC53463-250MG
2,3-Difluoro-4-formylbenzonitrile 96%
2,3-Difluoro-4-formylbenzonitrile
Formel: C₈H₃F₂NO
Molecular Weight: 167,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24684755
CAS-Nummer: 717883-41-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR