3'-Bromo-4',5'-difluoroacetophenone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(3-Bromo-4,5-difluorophenyl)ethanone
PC53468-1G
PC53468-250MG
APOSPC53468-1GEA
783
EUR
APOSPC53468-1G
APOSPC53468-250MG
3'-Bromo-4',5'-difluoroacetophenone
3'-Bromo-4',5'-difluoroacetophenone
Formel:
C₈H₅BrF₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD28737869 CAS-Nummer: 1805420-44-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...