3-(Trifluoromethylsulphonyl)benzonitrile

Lieferant: Apollo Scientific
Achtung

PC53472-1G PC53472-250MG
APOSPC53472-1GEA 579 EUR
APOSPC53472-1G APOSPC53472-250MG
3-(Trifluoromethylsulphonyl)benzonitrile
3-(Trifluoromethylsulphonyl)benzonitrile
Formel: C₈H₄F₃NO₂S
Molecular Weight: 235.183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25460328
CAS-Nummer: 1274903-99-9
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR