2,6-Difluoro-3-(trifluoromethyl)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

PC53476-1G PC53476-250MG
APOSPC53476-1GEA 1290 EUR
APOSPC53476-1G APOSPC53476-250MG
2,6-Difluoro-3-(trifluoromethyl)benzyl alcohol
2,6-Difluoro-3-(trifluoromethyl)benzyl alcohol
Formel: C₈H₅F₅O
Molecular Weight: 212,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30723302
CAS-Nummer: 1800231-22-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR