2,2,3,3-Tetrafluoro-6-iodo-1,4-benzodioxane

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2,3,3-Tetrafluoro-6-iodo-2,3-dihydro-1,4-benzodioxine

PC53484-1G PC53484-250MG
APOSPC53484-1GEA 783 EUR
APOSPC53484-1G APOSPC53484-250MG
2,2,3,3-Tetrafluoro-6-iodo-1,4-benzodioxane
2,2,3,3-Tetrafluoro-6-iodo-1,4-benzodioxane
Formel: C₈H₃F₄IO₂
Molecular Weight: 334,006 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30723346
CAS-Nummer: 2149597-65-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR