6-Amino-7-nitro-2,2,3,3-tetrafluoro-1,4-benzodioxane

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2,3,3-Tetrafluoro-7-nitro-2,3-dihydro-1,4-benzodioxin-6-amine

PC53490-1G PC53490-250MG
APOSPC53490-1GEA 574 EUR
APOSPC53490-1G APOSPC53490-250MG
6-Amino-7-nitro-2,2,3,3-tetrafluoro-1,4-benzodioxane
6-Amino-7-nitro-2,2,3,3-tetrafluoro-1,4-benzodioxane
Formel: C₈H₄F₄N₂O₄
Molecular Weight: 268,122 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30723440
CAS-Nummer: 2149597-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR