6,7-Diamino-2,2,3,3-tetrafluoro-1,4-benzodioxane 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,2,3,3-Tetrafluoro-2,3-dihydro-1,4-benzodioxine-6,7-diamine
PC53499-1G
PC53499-250MG
APOSPC53499-1GEA
841
EUR
APOSPC53499-1G
APOSPC53499-250MG
6,7-Diamino-2,2,3,3-tetrafluoro-1,4-benzodioxane 95%
6,7-Diamino-2,2,3,3-tetrafluoro-1,4-benzodioxane
Formel:
C₈H₆F₄N₂O₂ Molecular Weight: 238,139 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30723441 CAS-Nummer: 110968-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...