2-Amino-3-fluorobenzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

PC535007-1G PC535007-250MG
APOSPC535007-1GEA 157 EUR
APOSPC535007-1G APOSPC535007-250MG
2-Amino-3-fluorobenzaldehyde 95%
2-Amino-3-fluorobenzaldehyde
Formel: C₇H₆FNO
Molecular Weight: 139.127 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12923269
CAS-Nummer: 854538-94-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR