2,4-Difluoro-5-iodobenzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC53501-1G
PC53501-250MG
PC53501-5G
APOSPC53501-1GEA
319
EUR
APOSPC53501-1G
APOSPC53501-250MG
APOSPC53501-5G
2,4-Difluoro-5-iodobenzaldehyde 95%
2,4-Difluoro-5-iodobenzaldehyde
Formel:
C₇H₃F₂IO Lagertemperatur: Raumtemperatur |
MDL:
MFCD28789181 CAS-Nummer: 1097626-30-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...