6-Chloro-2-(difluoromethyl)-3-fluoropyridine 95+%

Lieferant: Apollo Scientific
Achtung

PC535022-100MG PC535022-250MG
APOSPC535022-100MGEA 186 EUR
APOSPC535022-100MG APOSPC535022-250MG
6-Chloro-2-(difluoromethyl)-3-fluoropyridine 95+%
6-Chloro-2-(difluoromethyl)-3-fluoropyridine
Formel: C₆H₃ClF₃N
Molecular Weight: 181,543 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD25478439
CAS-Nummer: 1805304-78-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR