2-Amino-5-(trifluoromethyl)pyridin-3-ol 95+%

Lieferant: Apollo Scientific
Achtung

PC535033-100MG PC535033-250MG
APOSPC535033-100MGEA 115 EUR
APOSPC535033-100MG APOSPC535033-250MG
2-Amino-5-(trifluoromethyl)pyridin-3-ol 95+%
2-Amino-5-(trifluoromethyl)pyridin-3-ol
Formel: C₆H₅F₃N₂O
Molecular Weight: 178.112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16606179
CAS-Nummer: 1227586-78-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR