2,6-Dibromo-4-fluoropyridine 95+%

Lieferant: Apollo Scientific
Achtung

PC535042-250MG PC535042-1G
APOSPC535042-250MGEA 120 EUR
APOSPC535042-250MG APOSPC535042-1G
2,6-Dibromo-4-fluoropyridine 95+%
2,6-Dibromo-4-fluoropyridine
Formel: C₅H₂Br₂FN
Molecular Weight: 254,882 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13185514
CAS-Nummer: 1214344-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR