4-Chloro-3-fluoro-2-methoxypyridine 95+%

Lieferant: Apollo Scientific
Achtung

PC535045-100MG PC535045-250MG
APOSPC535045-100MGEA 78.3 EUR
APOSPC535045-100MG APOSPC535045-250MG
4-Chloro-3-fluoro-2-methoxypyridine 95+%
4-Chloro-3-fluoro-2-methoxypyridine
Formel: C₆H₅ClFNO
Molecular Weight: 161,561 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16610381
CAS-Nummer: 1227572-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR