2-Fluoro-3-(methylthio)pyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC535047-1G
PC535047-250MG
APOSPC535047-1GEA
34.8
EUR
APOSPC535047-1G
APOSPC535047-250MG
2-Fluoro-3-(methylthio)pyridine 95%
2-Fluoro-3-(methylthio)pyridine
Formel:
C₆H₆FNS Molecular Weight: 143.182 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22543684 CAS-Nummer: 210992-56-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...