3,4-Difluoropyridin-2-amine 97%

Lieferant: Apollo Scientific
Achtung

PC535053-100MG PC535053-250MG
APOSPC535053-100MGEA 190 EUR
APOSPC535053-100MG APOSPC535053-250MG
3,4-Difluoropyridin-2-amine 97%
3,4-Difluoropyridin-2-amine
Formel: C₅H₄F₂N₂
Molecular Weight: 130.095 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28722270
CAS-Nummer: 1804379-06-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR