6-Fluorobenzo[b]thiophene 95+%

Lieferant: Apollo Scientific
Achtung

PC535065-1G PC535065-250MG
APOSPC535065-1GEA 178 EUR
APOSPC535065-1G APOSPC535065-250MG
6-Fluorobenzo[b]thiophene 95+%
6-Fluorobenzo[b]thiophene
Formel: C₈H₅FS
Molecular Weight: 152,189 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18207210
CAS-Nummer: 205055-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR