4-Fluoro-1H-indole-6-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC535068-100MG
PC535068-250MG
APOSPC535068-100MGEA
74.1
EUR
APOSPC535068-100MG
APOSPC535068-250MG
4-Fluoro-1H-indole-6-carboxylic acid 95%
4-Fluoro-1H-indole-6-carboxylic acid
Formel:
C₉H₆FNO₂ Molecular Weight: 179,148 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07781705 CAS-Nummer: 885520-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...