6-Fluoronaphthalen-2-ol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC535072-100MG
PC535072-250MG
APOSPC535072-100MGEA
72.5
EUR
APOSPC535072-100MG
APOSPC535072-250MG
6-Fluoronaphthalen-2-ol 95+%
6-Fluoronaphthalen-2-ol
Formel:
C₁₀H₇FO Molecular Weight: 162,16 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18412124 CAS-Nummer: 13101-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...