2,2,2-Trifluoro-1-(1H-pyrazol-4-yl)ethanone 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC535074-100MG
PC535074-250MG
APOSPC535074-100MGEA
128
EUR
APOSPC535074-100MG
APOSPC535074-250MG
2,2,2-Trifluoro-1-(1H-pyrazol-4-yl)ethanone 95+%
2,2,2-Trifluoro-1-(1H-pyrazol-4-yl)ethanone
Formel:
C₅H₃F₃N₂O Molecular Weight: 164,085 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18879656 CAS-Nummer: 161957-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...