4-Fluoro-1-methyl-1H-indole-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

PC535076-100MG PC535076-250MG
APOSPC535076-100MGEA 148 EUR
APOSPC535076-100MG APOSPC535076-250MG
4-Fluoro-1-methyl-1H-indole-2-carboxylic acid 97%
4-Fluoro-1-methyl-1H-indole-2-carboxylic acid
Formel: C₁₀H₈FNO₂
Molecular Weight: 193,174 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11869190
CAS-Nummer: 1146291-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR