4-Chloro-6-fluoroindan-1-one 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-6-Fluoroindan-1-one

PC535080-100MG PC535080-250MG
APOSPC535080-100MGEA 21.9 EUR
APOSPC535080-100MG APOSPC535080-250MG
4-Chloro-6-fluoroindan-1-one 98%
4-Chloro-6-fluoroindan-1-one
Formel: C₉H₆ClFO
Molecular Weight: 184.595 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09038173
CAS-Nummer: 166250-01-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR