6-Fluorochroman-8-carbaldehyde 95+%

Lieferant: Apollo Scientific
Achtung

PC535082-100MG PC535082-250MG
APOSPC535082-100MGEA 27.7 EUR
APOSPC535082-100MG APOSPC535082-250MG
6-Fluorochroman-8-carbaldehyde 95+%
6-Fluorochroman-8-carbaldehyde
Formel: C₁₀H₉FO₂
Molecular Weight: 180,176 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08236762
CAS-Nummer: 82060-91-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR