6-Bromo-4-fluoro-2,3-dihydro-1H-inden-1-one 95+%

Lieferant: Apollo Scientific
Achtung

PC535085-1G PC535085-250MG
APOSPC535085-1GEA 149 EUR
APOSPC535085-1G APOSPC535085-250MG
6-Bromo-4-fluoro-2,3-dihydro-1H-inden-1-one 95+%
6-Bromo-4-fluoro-2,3-dihydro-1H-inden-1-one
Formel: C₉H₆BrFO
Molecular Weight: 229,046 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14654077
CAS-Nummer: 881189-74-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR