2-Chloro-6-iodo-4-(trifluoromethoxy)aniline

Lieferant: Apollo Scientific
Achtung

PC53509-1G PC53509-250MG
APOSPC53509-1GEA 519 EUR
APOSPC53509-1G APOSPC53509-250MG
2-Chloro-6-iodo-4-(trifluoromethoxy)aniline
2-Chloro-6-iodo-4-(trifluoromethoxy)aniline
Formel: C₇H₄ClF₃INO
Molecular Weight: 337,465 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28753341
CAS-Nummer: 1805237-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR