(S)-6,7-Difluorochroman-4-amine 95%

Lieferant: Apollo Scientific
Achtung

PC535092-100MG PC535092-250MG
APOSPC535092-100MGEA 62.5 EUR
APOSPC535092-100MG APOSPC535092-250MG
(S)-6,7-Difluorochroman-4-amine 95%
(S)-6,7-Difluorochroman-4-amine
Formel: C₉H₉F₂NO
Molecular Weight: 185,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374041
CAS-Nummer: 1213126-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR