1-Ethanesulphonyl-4-fluorobenzene

Lieferant: Apollo Scientific
Achtung

PC53511-1G PC53511-250MG
APOSPC53511-1GEA 516 EUR
APOSPC53511-1G APOSPC53511-250MG
1-Ethanesulphonyl-4-fluorobenzene
1-Ethanesulphonyl-4-fluorobenzene
Formel: C₈H₉FO₂S
Molecular Weight: 188,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00865310
CAS-Nummer: 2924-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR