Methyl 5-amino-2-fluoro-4-hydroxybenzoate 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC53515-1G
PC53515-250MG
APOSPC53515-1GEA
862
EUR
APOSPC53515-1G
APOSPC53515-250MG
Methyl 5-amino-2-fluoro-4-hydroxybenzoate 95%
Methyl 5-amino-2-fluoro-4-hydroxybenzoate
Formel:
C₈H₈FNO₃ Molecular Weight: 185,152 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28554709 CAS-Nummer: 1781061-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...