2-Bromo-6-fluorobenzal fluoride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-(difluoromethyl)-3-fluorobenzene
PC53521-1G
PC53521-5G
APOSPC53521-1GEA
75.4
EUR
APOSPC53521-1G
APOSPC53521-5G
2-Bromo-6-fluorobenzal fluoride
2-Bromo-6-fluorobenzal fluoride
Formel:
C₇H₄BrF₃ Molecular Weight: 225,006 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14698533 CAS-Nummer: 1166820-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...