4-Amino-3-(4-fluorophenyl)butyric acid hydrochloride

Lieferant: Apollo Scientific
Achtung

PC53524-1G PC53524-250MG PC53524-50MG
APOSPC53524-1GEA 531 EUR
APOSPC53524-1G APOSPC53524-250MG APOSPC53524-50MG
4-Amino-3-(4-fluorophenyl)butyric acid hydrochloride
4-Amino-3-(4-fluorophenyl)butyric acid hydrochloride
Formel: C₁₀H₁₂FNO₂·ClH
Molecular Weight: 233,667 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29067351
CAS-Nummer: 1858241-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR