2-Amino-5-chloro-3-fluorobenzoic acid

Lieferant: Apollo Scientific
Achtung

PC53542-1G PC53542-250MG
APOSPC53542-1GEA 202 EUR
APOSPC53542-1G APOSPC53542-250MG
2-Amino-5-chloro-3-fluorobenzoic acid
2-Amino-5-chloro-3-fluorobenzoic acid
Formel: C₇H₅ClFNO₂
Molecular Weight: 189.572 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11193641
CAS-Nummer: 1028757-83-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR