3-(Cyanomethyl)-5-fluorobenzeneboronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-(Cyanomethyl)-5-fluorophenyl)boronic acid
PC53544-1G
PC53544-250MG
APOSPC53544-1GEA
556
EUR
APOSPC53544-1G
APOSPC53544-250MG
3-(Cyanomethyl)-5-fluorobenzeneboronic acid
3-(Cyanomethyl)-5-fluorobenzeneboronic acid
Formel:
C₈H₇BFNO₂ Molecular Weight: 178,956 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD27935531 CAS-Nummer: 1460307-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...