2,4-Difluoro-5-methylbenzenesulphonyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

PC53548-1G PC53548-250MG
APOSPC53548-1GEA 400 EUR
APOSPC53548-1G APOSPC53548-250MG
2,4-Difluoro-5-methylbenzenesulphonyl chloride 97%
2,4-Difluoro-5-methylbenzenesulphonyl chloride
Formel: C₇H₅ClF₂O₂S
Molecular Weight: 226.628 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22196689
CAS-Nummer: 1235407-51-8
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR