2-Amino-5-(difluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Difluoromethyl)pyridin-2-amine

PC53561-1G PC53561-250MG PC53561-500MG
APOSPC53561-1GEA 876 EUR
APOSPC53561-1G APOSPC53561-250MG APOSPC53561-500MG
2-Amino-5-(difluoromethyl)pyridine
2-Amino-5-(difluoromethyl)pyridine
Formel: C₆H₆F₂N₂
Molecular Weight: 144.122 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18256527
CAS-Nummer: 1211523-68-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR