2-Bromo-4-fluoro-3-methyl-6-nitrophenol

Lieferant: Apollo Scientific
Achtung

PC53564-1G PC53564-250MG
APOSPC53564-1GEA 480 EUR
APOSPC53564-1G APOSPC53564-250MG
2-Bromo-4-fluoro-3-methyl-6-nitrophenol
2-Bromo-4-fluoro-3-methyl-6-nitrophenol
Formel: C₇H₅BrFNO₃
Molecular Weight: 250,022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30729942
CAS-Nummer: 2055699-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR