3-Bromo-5-[(trifluoromethyl)sulphonyl]benzoic acid

Lieferant: Apollo Scientific
Achtung

PC53567-1G PC53567-250MG
APOSPC53567-1GEA 435 EUR
APOSPC53567-1G APOSPC53567-250MG
3-Bromo-5-[(trifluoromethyl)sulphonyl]benzoic acid
3-Bromo-5-[(trifluoromethyl)sulphonyl]benzoic acid
Formel: C₈H₄BrF₃O₄S
Molecular Weight: 333,079 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30729943
CAS-Nummer: 2149590-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR