5-Bromo-3-chloro-2-fluorobenzotrifluoride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-1-chloro-2-fluoro-3-(trifluoromethyl)benzene

PC53575-1G PC53575-250MG PC53575-5G
APOSPC53575-1GEA 43.5 EUR
APOSPC53575-1G APOSPC53575-250MG APOSPC53575-5G
5-Bromo-3-chloro-2-fluorobenzotrifluoride 95%
5-Bromo-3-chloro-2-fluorobenzotrifluoride
Formel: C₇H₂BrClF₄
Molecular Weight: 277,441 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09033157
CAS-Nummer: 914225-67-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR