3-Bromo-2-iodo-5-(trifluoromethoxy)aniline 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC53578-1G
PC53578-250MG
APOSPC53578-1GEA
638
EUR
APOSPC53578-1G
APOSPC53578-250MG
3-Bromo-2-iodo-5-(trifluoromethoxy)aniline 95%
3-Bromo-2-iodo-5-(trifluoromethoxy)aniline
Formel:
C₇H₄BrF₃INO Molecular Weight: 381,916 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28741480 CAS-Nummer: 1807261-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...