4-Chloro-6-fluoropyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
PC5358-1G
PC5358-250MG
APOSPC5358-1GEA
364
EUR
APOSPC5358-1G
APOSPC5358-250MG
4-Chloro-6-fluoropyrimidine
4-Chloro-6-fluoropyrimidine
Formel:
C₄H₂ClFN₂ Molecular Weight: 132,523 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD09870037 CAS-Nummer: 51422-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...