Methyl 2-bromo-4-(difluoromethyl)benzoate
Lieferant: Apollo Scientific
|
Achtung
|
PC53586-1G
PC53586-250MG
APOSPC53586-1GEA
567
EUR
APOSPC53586-1G
APOSPC53586-250MG
Methyl 2-bromo-4-(difluoromethyl)benzoate
Methyl 2-bromo-4-(difluoromethyl)benzoate
Formel:
C₉H₇BrF₂O₂ Molecular Weight: 265.051 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31726336 CAS-Nummer: 2091157-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...